Diethyl chlorophosphite, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5


M-Clarity™ quality level = MQ100

D91659-100G D91659-25G D91659-5G
SIALD91659-100GEA 390 GBP
SIALD91659-100G SIALD91659-25G SIALD91659-5G
Diethyl chlorophosphite, Sigma-Aldrich®
Diethyl chlorophosphite
Formula: C₄H₁₀ClO₂P
MW: 156.55 g/mol
MDL Number: MFCD00009074
CAS Number: 589-57-1
EINECS: 209-652-2
UN: 2920
ADR: 8 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR