4-Methylbenzoyl isothiocyanate

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

009518-1G 009518-5G
FLUO009518-1GEA 0 GBP
FLUO009518-1G FLUO009518-5G
4-Methylbenzoyl isothiocyanate
4-Methylbenzoyl isothiocyanate
Formula: C₉H₇NOS
MDL Number: MFCD00060380
CAS Number: 16794-68-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR