3-Methylbenzoyl isothiocyanate

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

009517-1G 009517-5G
FLUO009517-1GEA 0 GBP
FLUO009517-1G FLUO009517-5G
3-Methylbenzoyl isothiocyanate
3-Methylbenzoyl isothiocyanate
Formula: C₉H₇NOS
MDL Number: MFCD00060378
CAS Number: 28115-86-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR