4,4'-(Hexafluoroisopropylidene)diphenol

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 4,4′-(Hexafluoroisopropylidene)diphenol, 2,2-Bis(4-hydroxyphenyl)hexafluoropropane, 4,4'-(Perfluoropropane-2,2-diyl)diphenol

003139-100G 003139-250G
FLUO003139-100GEA 0 GBP
FLUO003139-100G FLUO003139-250G
4,4'-(Hexafluoroisopropylidene)diphenol
4,4'-(Hexafluoroisopropylidene)diphenol
Formula: C₁₅H₁₀F₆O₂
MW: 336.24 g/mol
MDL Number: MFCD00000439
CAS Number: 1478-61-1
EINECS: 216-036-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR