3-Nitrobenzoyl isothiocyanate

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

009547-1G 009547-5G
FLUO009547-1GEA 0 GBP
FLUO009547-1G FLUO009547-5G
3-Nitrobenzoyl isothiocyanate
3-Nitrobenzoyl isothiocyanate
Formula: C₈H₄N₂O₃S
MDL Number: MFCD00041247
CAS Number: 78225-78-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR