2-Chloro-6-fluorobenzyl mercaptan
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
004446-1G
004446-5G
FLUO004446-1GEA
0
GBP
FLUO004446-1G
FLUO004446-5G
2-Chloro-6-fluorobenzyl mercaptan
2-Chloro-6-fluorobenzyl mercaptan
Formula:
C₇H₆ClFS |
MDL Number:
MFCD00052473 CAS Number: 170924-52-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...