3-Chloro-4-fluorobenzyl mercaptan

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

024376-1G 024376-5G
FLUO024376-1GEA 0 GBP
FLUO024376-1G FLUO024376-5G
3-Chloro-4-fluorobenzyl mercaptan
3-Chloro-4-fluorobenzyl mercaptan
Formula: C₇H₆ClFS
MDL Number: MFCD06798035
CAS Number: 886498-16-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR