3-Chloro-2-fluorobenzyl mercaptan
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
024375-1G
024375-5G
24375-5G
FLUO024375-1GEA
0
GBP
FLUO024375-1G
FLUO024375-5G
FLUO24375-5G
3-Chloro-2-fluorobenzyl mercaptan
3-Chloro-2-fluorobenzyl mercaptan
Formula:
C₇H₆ClFS |
MDL Number:
MFCD06798034 CAS Number: 333743-31-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...