4-Bromo-2-fluorobenzyl mercaptan

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

024380-1G 024380-5G
FLUO024380-1GEA 0 GBP
FLUO024380-1G FLUO024380-5G
4-Bromo-2-fluorobenzyl mercaptan
4-Bromo-2-fluorobenzyl mercaptan
Formula: C₇H₆BrFS
CAS Number: 886498-29-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR