2-Amino-6-fluorobenzamide

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

010565-1G 010565-5G 010565-25G
FLUO010565-1GEA 0 GBP
FLUO010565-1G FLUO010565-5G FLUO010565-25G
2-Amino-6-fluorobenzamide
2-Amino-6-fluorobenzamide
Formula: C₇H₇FN₂O
MDL Number: MFCD00816881
CAS Number: 115643-59-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR