Mucochloric acid, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
2,3-Dichloromalealdehydic acid
M-Clarity™ quality level = MQ100
M89803-100G
M89803-500G
SIALM89803-100GEA
37.8
GBP
SIALM89803-100G
SIALM89803-500G
Mucochloric acid, Sigma-Aldrich®
Mucochloric acid
Formula:
C₄H₂Cl₂O₃ MW: 168.96 g/mol |
MDL Number:
MFCD00135465 CAS Number: 87-56-9 EINECS: 201-752-4 UN: 2923 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...