Mucochloric acid, (max. 4% H₂O) 99% (dry weight)

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

B24523.22 B24523.36
B24523.22EA 20.3 GBP
B24523.22 B24523.36
Mucochloric acid, (max. 4% H₂O) 99% (dry weight)
Mucochloric acid

Purity: 99% CAS No.: 87-56-9 Molecular Formula: C4H2Cl2O3 Formula Weight: 168.96 Flash Point: 186°(367°F) Melting Point: 125-128°(dried) Beilstein Reference: 1705641 Merck Reference: 14,6297 UN No.: 2923 MDL No.: MFCD00006966

Formula: C₄H₂Cl₂O₃
Melting Pt: 125…128 °C (anhydr.)
Density: 1.85
Flash Pt: 186 °C (367 °F)
Storage Temperature: Ambient
MDL Number: MFCD00135465
CAS Number: 87-56-9
EINECS: 201-752-4
UN: 2923
ADR: 8,III
Merck Index: 14,6297

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR