(4,5-Dimethoxy-2-nitrophenyl)acetic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2-(4,5-Dimethoxy-2-nitrophenyl)acetic acid

CA-4076-5G CA-4076-1G CA-4076-25G
COBBCA-4076-5GEA 63.31 GBP
COBBCA-4076-5G COBBCA-4076-1G COBBCA-4076-25G
(4,5-Dimethoxy-2-nitrophenyl)acetic acid
(4,5-Dimethoxy-2-nitrophenyl)acetic acid
Formula: C₁₀H₁₁NO₆
MW: 241.20 g/mol
MDL Number: MFCD00118466
CAS Number: 73357-18-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR