Ethyl 3,4-dichlorobenzoylformate

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Ethyl (3,4-dichlorophenyl)(oxo)acetate

023002-1G 023002-25G
FLUO023002-1GEA 0 GBP
FLUO023002-1G FLUO023002-25G
Ethyl 3,4-dichlorobenzoylformate
Ethyl 3,4-dichlorobenzoylformate
Formula: C₁₀H₈Cl₂O₃
MW: 247.08 g/mol
MDL Number: MFCD01934864
CAS Number: 34966-52-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR