Tris[2-(2-methoxyethoxy)ethyl]amine, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
Tris(2-(2-methoxyethoxy)ethyl)amine, Tris(3,6-dioxaheptyl)amine, Tris(dioxa-3,6-heptyl)amine
301248-100G
301248-500G
301248-5G
SIAL301248-100GEA
210
GBP
SIAL301248-100G
SIAL301248-500G
SIAL301248-5G
Tris[2-(2-methoxyethoxy)ethyl]amine, Sigma-Aldrich®
Tris[2-(2-methoxyethoxy)ethyl]amine
Formula:
C₁₅H₃₃NO₆ MW: 323.43 g/mol |
MDL Number:
MFCD00010748 CAS Number: 70384-51-9 EINECS: 274-590-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...