4-Fluorophenyloxamic acid ethyl ester

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Ethyl 2-((4-fluorophenyl)amino)-2-oxoacetate, Ethyl 2-(4-fluoroanilino)-2-oxoacetate

010095-1G 010095-10G
FLUO010095-1GEA 0 GBP
FLUO010095-1G FLUO010095-10G
4-Fluorophenyloxamic acid ethyl ester
4-Fluorophenyloxamic acid ethyl ester
Formula: C₁₀H₁₀FNO₃
MW: 211.1897 g/mol
MDL Number: MFCD00119045
CAS Number: 69065-91-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR