L(-)-Malic acid disodium salt, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
M9138-100G
M9138-25G
M9138-5G
SIALM9138-100GEA
512
GBP
SIALM9138-100G
SIALM9138-25G
SIALM9138-5G
L(-)-Malic acid disodium salt, Sigma-Aldrich®
L(-)-Malic acid disodium salt
Formula:
C₄H₄Na₂O₅ |
CAS Number:
138-09-0 EINECS: 205-313-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...