1-(Trimethylsilyl)-1,2,4-triazole 95%
Supplier: Thermo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
Synonyms:
1-(Trimethylsilyl)-1H-1,2,4-triazole
B21105.06
B21105.14
B21105.06EA
69.6
GBP
B21105.06
B21105.14
1-(Trimethylsilyl)-1,2,4-triazole 95%
1-(Trimethylsilyl)-1,2,4-triazole
Purity: 98%
CAS No.: 18293-54-4
Molecular Formula: C5H11N3Si
Formula Weight: 141.25
Flash Point: -5°(23°F)
Refractive Index: 1.4610
Boiling Point: 73-75°/12mm
UN No.: 1993
MDL No.: MFCD00046070
Formula:
C₅H₁₁N₃Si MW: 141.25 g/mol Boiling Pt: 73…75 °C (12 mmHg) Density: 0.98 Flash Pt: –5 °C (23 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00046070 CAS Number: 18293-54-4 EINECS: 242-170-0 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...