4-Bromo-2-ethyl-1-iodobenzene

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

024167-25G 024167-5G
FLUO024167-25GEA 0 GBP
FLUO024167-25G FLUO024167-5G
4-Bromo-2-ethyl-1-iodobenzene
4-Bromo-2-ethyl-1-iodobenzene
Formula: C₈H₈BrI
MW: 310.97 g/mol
MDL Number: MFCD00221458
CAS Number: 175278-30-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR