1-Chloro-2-fluoro-4-iodobenzene
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
4-Chloro-3-fluoroiodobenzene
018374-5G
018374-25G
FLUO018374-5GEA
0
GBP
FLUO018374-5G
FLUO018374-25G
1-Chloro-2-fluoro-4-iodobenzene
1-Chloro-2-fluoro-4-iodobenzene
Formula:
C₆H₃ClFI |
MDL Number:
MFCD00042580 CAS Number: 202982-67-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...