5-Methylpyrazine-2-carboxylic acid
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
019499-25G
019499-100G
FLUO019499-25GEA
0
GBP
FLUO019499-25G
FLUO019499-100G
5-Methylpyrazine-2-carboxylic acid
5-Methylpyrazine-2-carboxylic acid
Formula:
C₆H₆N₂O₂ |
MDL Number:
MFCD00068241 CAS Number: 5521-55-1 EINECS: 413-260-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...