2-Chloro-6-(propylamino)pyrazine

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

HC-6265-5G HC-6265-1G
COBBHC-6265-5GEA 0 GBP
COBBHC-6265-5G COBBHC-6265-1G
2-Chloro-6-(propylamino)pyrazine
2-Chloro-6-(propylamino)pyrazine
Formula: C₇H₁₀ClN₃
MDL Number: MFCD09801000
CAS Number: 951884-52-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR