2-Chloro-6-(ethylamino)pyrazine

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

HC-6373-1G HC-6373-5G
COBBHC-6373-1GEA 0 GBP
COBBHC-6373-1G COBBHC-6373-5G
2-Chloro-6-(ethylamino)pyrazine
2-Chloro-6-(ethylamino)pyrazine
Formula: C₈H₁₂ClN₃
MDL Number: MFCD09878382
CAS Number: 957065-84-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR