2-Amino-5-bromopyrazine

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

PY-7820-25G PY-7820-5G
COBBPY-7820-25GEA 0 GBP
COBBPY-7820-25G COBBPY-7820-5G
2-Amino-5-bromopyrazine
2-Amino-5-bromopyrazine
Formula: C₄H₄BrN₃
MDL Number: MFCD00235015
CAS Number: 59489-71-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR