Chromium (III) sulphate hydrate, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5


M-Clarity™ quality level = MQ200

342432-100G 342432-500G
SIAL342432-100GEA 112 GBP
SIAL342432-100G SIAL342432-500G
Chromium (III) sulphate hydrate, Sigma-Aldrich®
Chromium (III) sulphate hydrate
Formula: Cr₂O₁₂S₃·H₂O
MDL Number: MFCD00150215
CAS Number: 15244-38-9
EINECS: 233-253-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR