N,O-Dimethylhydroxylamine hydrochloride

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

023913-1KG 023913-100G 023913-25G
FLUO023913-1KGEA 0 GBP
FLUO023913-1KG FLUO023913-100G FLUO023913-25G
N,O-Dimethylhydroxylamine hydrochloride
N,O-Dimethylhydroxylamine hydrochloride
Formula: C₂H₈ClNO
MDL Number: MFCD00012485
CAS Number: 6638-79-5
EINECS: 229-642-1

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR