1,4,7-Trimethyl-1,4,7-triazacyclononane ≥97%

Supplier: Strem Chemicals, Inc.
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 1,4,7-Trimethyl-1,4,7-triazonane

07-2750-2G 07-2750-500MG
STRE07-2750-2GEA 537.36 GBP
STRE07-2750-2G STRE07-2750-500MG
1,4,7-Trimethyl-1,4,7-triazacyclononane ≥97%
1,4,7-Trimethyl-1,4,7-triazacyclononane
Formula: C₉H₂₁N₃
MW: 171.28 g/mol
Density: 0.884
Flash Pt: 155 °F
MDL Number: MFCD00012359
CAS Number: 96556-05-7
UN: 3267
ADR: 8 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR