Diethyl mesoxalate (diethyl ketomalonate)
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
Diethyl 2-oxomalonate
005421-5G
005421-25G
005421-100G
FLUO005421-5GEA
0
GBP
FLUO005421-5G
FLUO005421-25G
FLUO005421-100G
Diethyl mesoxalate (diethyl ketomalonate)
Diethyl mesoxalate (Diethyl ketomalonate)
Formula:
C₇H₁₀O₅ |
MDL Number:
MFCD00009121 CAS Number: 609-09-6 EINECS: 210-176-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...