1,4-Bis(1,1,2,2-tetrafluoroethoxy)benzene

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

010421-10G 010421-25G
FLUO010421-10GEA 0 GBP
FLUO010421-10G FLUO010421-25G
1,4-Bis(1,1,2,2-tetrafluoroethoxy)benzene
1,4-Bis(1,1,2,2-tetrafluoroethoxy)benzene
Formula: C₁₀H₆F₈O₂
CAS Number: 3832-65-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR