1,2-Bis(1,1,2,2-tetrafluoroethoxy)benzene

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

010419-25G 010419-5G
FLUO010419-25GEA 0 GBP
FLUO010419-25G FLUO010419-5G
1,2-Bis(1,1,2,2-tetrafluoroethoxy)benzene
1,2-Bis(1,1,2,2-tetrafluoroethoxy)benzene
Formula: C₁₀H₆F₈O₂
MDL Number: MFCD03424468
CAS Number: 4063-48-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR