7-Bromooxindole
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
IN-0092-5G
IN-0092-1G
COBBIN-0092-5GEA
0
GBP
COBBIN-0092-5G
COBBIN-0092-1G
7-Bromooxindole
7-Bromooxindole
Formula:
C₈H₆BrNO MW: 212.0433 g/mol |
MDL Number:
MFCD02179613 CAS Number: 320734-35-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...