4-Bromo-1-fluoro-2-iodobenzene

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

OS-7690-5G OS-7690-25G OS-7690-100G
COBBOS-7690-5GEA 0 GBP
COBBOS-7690-5G COBBOS-7690-25G COBBOS-7690-100G
4-Bromo-1-fluoro-2-iodobenzene
4-Bromo-1-fluoro-2-iodobenzene
Formula: C₆H₃BrFI
MW: 300.9000 g/mol
MDL Number: MFCD07368751
CAS Number: 116272-41-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR