Carmine

Supplier: Serva
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

16180.01 16180.02
SERA16180.01EA 0 GBP
SERA16180.01 SERA16180.02
Carmine
Carmine
Formula: C₂₂H₂₀O₁₃
MDL Number: MFCD00167028
CAS Number: 1390-65-4
EINECS: 215-724-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR