Methyl-β-D-glucopyranoside, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
Methyl β-D-glucopyranoside, (2R,3S,4S,5R,6R)-2-(Hydroxymethyl)-6-methoxytetrahydro-2H-pyran-3,4,5-triol
M-Clarity™ quality level = MQ200
M0779-100G
M0779-5G
SIALM0779-100GEA
1990
GBP
SIALM0779-100G
SIALM0779-5G
Methyl-β-D-glucopyranoside, Sigma-Aldrich®
Methyl β-D-glucopyranoside
Formula:
C₇H₁₄O₆ MW: 194.1825 g/mol |
MDL Number:
MFCD00006602 CAS Number: 709-50-2 EINECS: 211-909-9 |
Learn more
![](/m-uk.vwr.com/en_GB/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...