2-Iodo-4-(trifluoromethoxy)aniline

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

023865-25G 023865-5G
FLUO023865-25GEA 0 GBP
FLUO023865-25G FLUO023865-5G
2-Iodo-4-(trifluoromethoxy)aniline
2-Iodo-4-(trifluoromethoxy)aniline
Formula: C₇H₅F₃INO
MDL Number: MFCD04972994
CAS Number: 845866-79-7
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR