4-(Trifluoromethyl)benzyl mercaptan

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

024355-1G 024355-5G
FLUO024355-1GEA 0 GBP
FLUO024355-1G FLUO024355-5G
4-(Trifluoromethyl)benzyl mercaptan
4-(Trifluoromethyl)benzyl mercaptan
Formula: C₈H₇F₃S
MDL Number: MFCD06798015
CAS Number: 108499-24-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR