1-Bromo-2-chloro-4-iodobenzene

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

021409-5G 021409-25G
FLUO021409-5GEA 0 GBP
FLUO021409-5G FLUO021409-25G
1-Bromo-2-chloro-4-iodobenzene
1-Bromo-2-chloro-4-iodobenzene
Formula: C₆H₃BrCl
MDL Number: MFCD04973904
CAS Number: 535934-25-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR