Bis(pentamethylcyclopentadienyl)nickel(II)

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: Bis(pentamethylcyclopentadienyl)nickel, Decamethylnickelocene

39238.03 39238.06 39238.MD
39238.03EA 163 GBP
39238.03 39238.06 39238.MD
Bis(pentamethylcyclopentadienyl)nickel(II)
Bis(pentamethylcyclopentadienyl)nickel (II)

CAS No.: 74507-63-4 Molecular Formula: C20H30Ni Formula Weight: 329.17 Physical Form: Crystalline, Packaged in ampoules Sensitivity: Air and Moisture Sensitive

Formula: C₂₀H₃₀Ni
Storage Temperature: Ambient
MDL Number: MFCD00058704
CAS Number: 74507-63-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR