Tripalmitin (glycerol tripalmitate), Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Propane-1,2,3-triyl tripalmitate, Tripalmitin, Glycerol tripalmitate

T5888-1G T5888-5G T5888-100MG T5888-500MG
SIALT5888-1GEA 195 GBP
SIALT5888-1G SIALT5888-5G SIALT5888-100MG SIALT5888-500MG
Tripalmitin (glycerol tripalmitate), Sigma-Aldrich®
Tripalmitin (Glycerol tripalmitate)
Formula: C₅₁H₉₈O₆
MDL Number: MFCD00008995
CAS Number: 555-44-2
EINECS: 209-098-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR