2-Bromo-α,α,α,6-tetrafluorotoluene

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 1-Bromo-3-fluoro-2-(trifluoromethyl)benzene, 2-Bromo-6-fluorobenzotrifluoride

OS-2101-25G OS-2101-100G
COBBOS-2101-25GEA 0 GBP
COBBOS-2101-25G COBBOS-2101-100G
2-Bromo-α,α,α,6-tetrafluorotoluene
2-Bromo-α,α,α,6-tetrafluorotoluene
Formula: C₇H₃BrF₄
MDL Number: MFCD01631569
CAS Number: 261951-85-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR