2-Bromo-4-(trifluoromethyl)anisole

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

010726-5G 010726-25G
FLUO010726-5GEA 0 GBP
FLUO010726-5G FLUO010726-25G
2-Bromo-4-(trifluoromethyl)anisole
2-Bromo-4-(trifluoromethyl)anisole
Formula: C₈H₆BrF₃O
MDL Number: MFCD03789249
CAS Number: 402-10-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR