2-Propyl-1,3,2-benzodioxaborole (β-propylocatecholborane)

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

BB-8310-1G BB-8310-5G
COBBBB-8310-1GEA 0 GBP
COBBBB-8310-1G COBBBB-8310-5G
2-Propyl-1,3,2-benzodioxaborole (β-propylocatecholborane)
2-Propyl-1,3,2-benzodioxaborole (β-Propylocatecholborane)
Formula: C₉H₁₁BO₂
MDL Number: MFCD00799577
CAS Number: 40218-49-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR