2-(4-Bromobutyl)-1,3,2-benzodioxaborole (β-propylocatecholborane)
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
2-(4-Bromobutyl)-1,3,2-benzodioxaborole
BB-8343-5G
BB-8343-1G
COBBBB-8343-5GEA
0
GBP
COBBBB-8343-5G
COBBBB-8343-1G
2-(4-Bromobutyl)-1,3,2-benzodioxaborole (β-propylocatecholborane)
2-(4-Bromobutyl)-1,3,2-benzodioxaborole (β-Propylocatecholborane)
Formula:
C₁₀H₁₂BBrO₂ |
MDL Number:
MFCD01863716 CAS Number: 142172-51-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...