2-Bromo-1,3,2-benzodioxaborole (β-bromocatecholborane), Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2-Bromo-1,3,2-benzodioxaborole, B-Bromocatecholborane

M-Clarity™ quality level = MQ100

304166-10G 304166-50G
SIAL304166-10GEA 143 GBP
SIAL304166-10G SIAL304166-50G
2-Bromo-1,3,2-benzodioxaborole (β-bromocatecholborane), Sigma-Aldrich®
2-Bromo-1,3,2-benzodioxaborole (β-Bromocatecholborane)
Formula: C₆H₄BBrO₂
MDL Number: MFCD00013341
CAS Number: 51901-85-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR