2-Methoxycinnamaldehyde, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
M-Clarity™ quality level = MQ300
W318108-100G-K
W318108-5KG-K
SIALW318108-100G-KEA
90.5
GBP
SIALW318108-100G-K
SIALW318108-5KG-K
2-Methoxycinnamaldehyde, Sigma-Aldrich®
2-Methoxycinnamaldehyde
Formula:
C₁₀H₁₀O₂ MW: 162.1852 g/mol |
MDL Number:
MFCD00007001 CAS Number: 1504-74-1 EINECS: 216-131-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...