(6-Bromo-2-pyridinyl)methanol, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
(6-Bromopyridin-2-yl)methanol, 2-Bromo-6-pyridinemethanol, 2-Bromo-6-(hydroxymethyl)pyridine
M-Clarity™ quality level = MQ100
554146-5G
554146-1G
SIAL554146-5GEA
109
GBP
SIAL554146-5G
SIAL554146-1G
(6-Bromo-2-pyridinyl)methanol, Sigma-Aldrich®
(6-Bromo-2-pyridinyl)methanol
Formula:
C₆H₆BrNO MW: 188.02 g/mol |
MDL Number:
MFCD04039884 CAS Number: 33674-96-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...