4-Bromotetrahydropyran

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

OT-8120-5G OT-8120-25G OT-8120-1G
COBBOT-8120-5GEA 0 GBP
COBBOT-8120-5G COBBOT-8120-25G COBBOT-8120-1G
4-Bromotetrahydropyran
4-Bromotetrahydropyran
Formula: C₅H₉BrO
MDL Number: MFCD07371501
CAS Number: 25637-16-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR