4-Chloro-2,8-dimethylquinoline
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
HC-2866-5G
HC-2866-1G
COBBHC-2866-5GEA
0
GBP
COBBHC-2866-5G
COBBHC-2866-1G
4-Chloro-2,8-dimethylquinoline
4-Chloro-2,8-dimethylquinoline
Formula:
C₁₁H₁₀ClN |
MDL Number:
MFCD00272352 CAS Number: 32314-39-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...