2-Chloro-6,8-dimethylquinoline

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

PY-1306-5G PY-1306-1G
COBBPY-1306-5GEA 0 GBP
COBBPY-1306-5G COBBPY-1306-1G
2-Chloro-6,8-dimethylquinoline
2-Chloro-6,8-dimethylquinoline
Formula: C₁₁H₁₀ClN
MDL Number: MFCD15143624
CAS Number: 139719-23-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR