8-Quinolinesulphonyl chloride, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5


M-Clarity™ quality level = MQ100

Q1506-10G Q1506-5G
SIALQ1506-10GEA 110 GBP
SIALQ1506-10G SIALQ1506-5G
8-Quinolinesulphonyl chloride, Sigma-Aldrich®
8-Quinolinesulphonyl chloride
Formula: C₉H₆ClNO₂S
MDL Number: MFCD00006808
CAS Number: 18704-37-5
EINECS: 242-515-5
UN: 3261
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR